Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101483
Description
Antibody detects endogenous levels of total CLIP1.
CLIP170 was initially identified as a new type of intermediate filament associated protein that is highly expressed in Reed-Sternberg cells, the tumoral cells diagnostic for Hodgkin's disease. Later experiments showed that it is located at microtubule plus ends and is required for the binding of endocytic carrier vesicles. CLIP170 has also been suggested to act with LIS1, a protein implicated in brain development, to regulate dynein/dynactin binding microtubules. Other studies suggest that CLIP170 can influence the formation of lamellipodia and cell invasion by invasive breast cancer cells by regulating the release of kinesin and IQGAP1 from a complex of those proteins, CLIP170 and Rac1. At least two isoforms of CLIP170 are known to exist.
Specifications
CLIP1 | |
Polyclonal | |
Unconjugated | |
CLIP1 | |
1110007I12Rik; 4631429H07Rik; AV017631; C81039; CAP-Gly domain containing linker protein 1; CAP-Gly domain-containing linker protein 1; CLIP; Clip 170; Clip1; CLIP170; CLIP-170; Clip50; Cyln1; cytoplasmic linker protein 1; Cytoplasmic linker protein 170; Cytoplasmic linker protein 170 alpha-2; cytoplasmic linker protein 50; cytoplasmic linker protein CLIP-170; Kiaa4046; MGC131604; mKIAA4046; Reed-Steinberg cell-espressed intermediate filament-associated protein; Reed-Sternberg intermediate filament-associated protein; Restin; restin (Reed-Steinberg cell-espressed intermediate filament-associated protein); restin (Reed-Steinberg cell-expressed intermediate filament-associated protein); Rsn | |
Rabbit | |
Affinity chromatography | |
RUO | |
56430, 6249, 65201 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P30622, Q922J3, Q9JK25 | |
CLIP1 | |
A synthesized peptide derived from human CLIP1(Accession P30622), corresponding to amino acid residues C1384-D1434. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction