Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLIP1 Recombinant Rabbit Monoclonal Antibody (JE63-82)
GREENER_CHOICE

Catalog No. PIMA544803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544803 Supplier Invitrogen™ Supplier No. MA544803
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CLIP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

CLIP170 was initially identified as a new type of intermediate filament associated protein that is highly expressed in Reed-Sternberg cells, the tumoral cells diagnostic for Hodgkin's disease. Later experiments showed that it is located at microtubule plus ends and is required for the binding of endocytic carrier vesicles. CLIP170 has also been suggested to act with LIS1, a protein implicated in brain development, to regulate dynein/dynactin binding microtubules. Other studies suggest that CLIP170 can influence the formation of lamellipodia and cell invasion by invasive breast cancer cells by regulating the release of kinesin and IQGAP1 from a complex of those proteins, CLIP170 and Rac1. At least two isoforms of CLIP170 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE63-82
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CLIP1
Gene Accession No. P30622
Gene Alias 1110007I12Rik; 4631429H07Rik; AV017631; C81039; CAP-Gly domain containing linker protein 1; CAP-Gly domain-containing linker protein 1; CLIP; Clip 170; Clip1; CLIP170; CLIP-170; Clip50; Cyln1; cytoplasmic linker protein 1; Cytoplasmic linker protein 170; Cytoplasmic linker protein 170 alpha-2; cytoplasmic linker protein 50; cytoplasmic linker protein CLIP-170; Kiaa4046; MGC131604; mKIAA4046; Reed-Steinberg cell-espressed intermediate filament-associated protein; Reed-Sternberg intermediate filament-associated protein; Restin; restin (Reed-Steinberg cell-espressed intermediate filament-associated protein); restin (Reed-Steinberg cell-expressed intermediate filament-associated protein); Rsn
Gene Symbols CLIP1
Host Species Rabbit
Immunogen Synthetic peptide within human CLIP170 aa 1,389-1,438/1,438.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6249
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.