Learn More
CLIP170 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303345AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
CLIP170 was initially identified as a new type of intermediate filament associated protein that is highly expressed in Reed-Sternberg cells, the tumoral cells diagnostic for Hodgkin′s disease. Later experiments showed that it is located at microtubule plus ends and is required for the binding of endocytic carrier vesicles. CLIP170 has also been suggested to act with LIS1, a protein implicated in brain development, to regulate dynein/dynactin binding microtubules. Other studies suggest that CLIP170 can influence the formation of lamellipodia and cell invasion by invasive breast cancer cells by regulating the release of kinesin and IQGAP1 from a complex of those proteins, CLIP170 and Rac1. At least two isoforms of CLIP170 are known to exist.Specifications
CLIP170 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P30622 | |
CLIP1 | |
Between 1388 and 1438 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
CLIP1 | |
CLIP1; CAP-GLY domain containing linker protein 1; CLIP; CLIP-170; CLIP170; CYLN1; MGC131604; RSN; Reed-Steinberg cell-expressed intermediate filament-associated protein; cytoplasmic linker 1; restin; restin (Reed-Steinberg cell-expressed intermediate filament-associated protein) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6249 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.