Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113370
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113370 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113370 Supplier Invitrogen™ Supplier No. PA5113370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLIP2 Polyclonal Antibody for ELISA, ICC/IF, Western Blot

The protein encoded by this gene belongs to the family of cytoplasmic linker proteins, which have been proposed to mediate the interaction between specific membranous organelles and microtubules. This protein was found to associate with both microtubules and an organelle called the dendritic lamellar body. This gene is hemizygously deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIP2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CLIP2
Gene Accession No. O55156, Q9UDT6, Q9Z0H8
Gene Alias B230327O20; CAP-Gly domain containing linker protein 2; CAP-Gly domain-containing linker protein 2; CLIP; Clip1; CLIP-115; Clip2; Cyln2; cytoplasmic linker 2; cytoplasmic linker protein 1, 115 kDa; cytoplasmic linker protein 115; Cytoplasmic linker protein 2; Kiaa0291; mKIAA0291; testicular tissue protein Li 40; WBSCR3; WBSCR4; williams-Beuren syndrome chromosomal region 3 protein; Williams-Beuren syndrome chromosomal region 4 protein; Williams-Beuren syndrome chromosome region 3; Williams-Beuren syndrome chromosome region 4; WSCR3; WSCR4
Gene Symbols CLIP2
Host Species Rabbit
Immunogen Recombinant Human CAP-Gly domain-containing linker protein 2 protein (101-217AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269713, 29264, 7461
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.