Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLIP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570093 Supplier Invitrogen™ Supplier No. PA570093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

Functions as a cytoplasmic linker protein. Involved in TGN-endosome dynamics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIP3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CLIP3
Gene Accession No. B9EHT4
Gene Alias 1500005P14Rik; AI844915; CAP-GLY domain containing linker protein 3; CAP-Gly domain-containing linker protein 3; clip-170 related protein 59 kDa; CLIP-170-related 59 kDa protein; Clip3; CLIPR59; Clipr-59; Cytoplasmic linker protein 170-related 59 kDa protein; restin-like 1; RGD1306245; RSNL1
Gene Symbols CLIP3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human CLIP3 (aa 9-58).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76686
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.