Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLIP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584281
Description
Immunogen sequence: SRVQRVTDSL DTLSEISSNK QNHSYPGFRR SFSTTSASSQ KEINRRNAFS KSKAALRRSW SSTPTAGGIE GSVKLHEGSQ VLLTSSNEMG TV.
CLIP4 includes 4 isoforms of the human protein that are produced by alternative splicing.
Specifications
CLIP4 | |
Polyclonal | |
Unconjugated | |
CLIP4 | |
1700024K14Rik; 1700074B05Rik; 4833417L20Rik; 5830409B12Rik; CAP-Gly domain containing linker protein family member 4; CAP-GLY domain containing linker protein family, member 4; CAP-Gly domain-containing linker protein 4; Clip4; restin-like 2; restin-like protein 2; RGD1311049; RSNL2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79745 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N3C7 | |
CLIP4 | |
Recombinant protein corresponding to Human CLIP4. Recombinant protein control fragment (Product #RP-105037). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction