Learn More
Promega CLK1 Kinase Enzyme System, 10microg
Catalog No. PRV4056 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4056 10 μg
1 options
Catalog No. PRV4056 Supplier Promega Supplier No. V4056
Recombinant human CLK1 (129-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK1, also known as STY, is a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases.
  • Easily Screen and Profile CLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C