Learn More
Promega CLK1 Kinase Enzyme System, 1mg
Catalog No. PRV6288 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6288 1 mg
1 options
Catalog No. PRV6288 Supplier Promega Supplier No. V6288
Add to Cart
Add to Cart
Request a Quote
Recombinant human CLK1 (129-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK1, also known as STY, is a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases.
  • Easily Screen and Profile CLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C