Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101117 Supplier Invitrogen™ Supplier No. PA5101117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CLK1.

CLK1 is a dual specificity kinase that phosphorylates PTP-1B and members of the SR family of splicing factors. It is thought to be involved in neuronal differentiation and mRNA splicing. Therefore, the encoded protein may play an indirect role in governing splice site selection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLK1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLK1
Gene Accession No. P22518, P49759
Gene Alias CDC like kinase 1; CDC28/CDC2-like kinase; CDC-like kinase 1; CLK; CLK/STY; CLK1; Dual specificity protein kinase CLK1; Protein kinase CLK1; protein kinase STY; protein tyrosine kinase STY; Sty
Gene Symbols CLK1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CLK1(Accession P49759), corresponding to amino acid residues K65-S115.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1195, 12747
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.