Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLK1 Polyclonal Antibody

Catalog No. PIPA528653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528653 Supplier Invitrogen™ Supplier No. PA528653

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Mouse (94%), Rat (94%), Xenopus laevis (80%), Pig (97%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CLK1 is a dual specificity kinase that phosphorylates PTP-1B and members of the SR family of splicing factors. It is thought to be involved in neuronal differentiation and mRNA splicing. Therefore, the encoded protein may play an indirect role in governing splice site selection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLK1
Applications Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CLK1
Gene Accession No. P49759
Gene Alias CDC like kinase 1; CDC28/CDC2-like kinase; CDC-like kinase 1; CLK; CLK/STY; CLK1; Dual specificity protein kinase CLK1; Protein kinase CLK1; protein kinase STY; protein tyrosine kinase STY; Sty
Gene Symbols CLK1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CLK1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1195
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.