Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111488 Supplier Invitrogen™ Supplier No. PA5111488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRVYDRRYCG SYRRNDYSRD RGDAYYDTDY RHSYEYQREN SSYRSQRSSR RKHRR Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 91%.

CLK2 is a serine/threonine kinase that phosphorylates serine/arginine-rich proteins as related to controlling RNA splicing. It is also thought to link cell cycle progression, apoptosis, and telomere length regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLK2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLK2
Gene Accession No. P49760
Gene Alias AU041688; CDC like kinase 2; CDC-like kinase 2; CLK kinase; Clk2; Clk2-Scamp3 protein; dual specificity protein kinase CLK2; EC 2.7.12.1; hCLK2; kinase CLK2; Protein kinase CLK2; Tu52
Gene Symbols CLK2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CLK2. Recombinant protein control fragment (Product #RP-106943).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1196
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.