Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 110921AP150UL

Description
This gene encodes a member of the CLK family of dual specificity protein kinases. CLK family members have been shown to interact with, and phosphorylate, serine- and arginine-rich (SR) proteins of the spliceosomal complex, which is a part of the regulatory mechanism that enables the SR proteins to control RNA splicing. Note that this gene is distinct from TELO2 gene (GeneID:9894), which shares CLK2 and hCLK2 symbol aliases in common with this gene, but encodes a protein that is involved in telomere length regulation.Specifications
CLK2 | |
Polyclonal | |
Unconjugated | |
CLK2 | |
CDC like kinase 2, CLK2, hCLK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1196, 12748, 365842 | |
-20°C | |
Liquid |
ELISA, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35491, P49760 | |
Clk2 | |
CLK2 Fusion Protein Ag1549 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction