Learn More
Promega CLK3 Kinase Enzyme System, 1mg
Catalog No. PRV6289 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6289 1 mg
1 options
Catalog No. PRV6289 Supplier Promega Supplier No. V6289
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human CLK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CLK3, also known as CDC-like kinase 3, encodes a serine/threonine-type protein kinase with a non-conserved N-terminal domain.
  • Easily Screen and Profile CLK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C