Learn More
Invitrogen™ CLK3 Monoclonal Antibody (1H2)

Description
Peptide Sequence: YPSRREPPPR RSRSRSHDRL PYQRRYRERR DSDTYRCEER SPSFGEDYYG PSRSRHRRRS RERGPYRTRK HAHHCHKRRT RSCSSASSRS QQSSKRSSRS V.
Specifications
Specifications
| Antigen | CLK3 |
| Applications | ELISA, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 1H2 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CLK3 |
| Gene Accession No. | P49761, Q63117 |
| Gene Alias | AI256811; CDC like kinase 3; cdc2/CDC28-like protein kinase 3; CDC-like kinase 3; Clk3; Dual specificity protein kinase CLK3; PHCLK3; PHCLK3/152; Protein kinase CLK3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.