Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517289
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517289 Supplier Thermo Scientific Supplier No. PA517289
Add to Cart
Add to Cart

CLK3 Polyclonal Antibody

CLK3 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen CLK3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 80 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene CLK3
Gene Accession No. O35492, P49761, Q63117
Gene Alias AI256811; CDC like kinase 3; cdc2/CDC28-like protein kinase 3; CDC-like kinase 3; Clk3; Dual specificity protein kinase CLK3; PHCLK3; PHCLK3/152; Protein kinase CLK3
Gene Symbols CLK3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Tyr209 of human CLK3 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102414, 1198, 171305
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.