Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLK3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988114
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8114 Supplier Bioss Supplier No. BS7908R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLK3 encodes a protein belonging to the serine/threonine type protein kinase family. This protein is a nuclear dual-specificity kinase that regulates the intranuclear distribution of the serine/arginine-rich (SR) family of splicing factors. Two transcript variants encoding different isoforms have been found for this gene. Related pseudogenes are located on chromosomes 1 and 9.

Specifications

Antigen CLK3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CLK3
Gene Accession No. P49761
Gene Alias AI256811; CDC like kinase 3; cdc2/CDC28-like protein kinase 3; CDC-like kinase 3; CLK3; Dual specificity protein kinase CLK3; PHCLK3; PHCLK3/152; Protein kinase CLK3
Gene Symbols CLK3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CLK3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1198
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.