Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLLD6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989197
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9197 Supplier Bioss Supplier No. BS9611R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C13orf1, also named as SPRYD7 and CLLD6, is involved in hematopoiesis via NOTCH signaling. C13orf1 plays an important role in the pathogenesis of B-CLL. There are isoforms of C13orf1 with MW 15-17 kDa and 19-22 kDa.

Specifications

Antigen CLLD6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Spryd7
Gene Accession No. Q5W111
Gene Alias 6330409N04Rik; C13orf1; Chronic lymphocytic leukemia deletion region gene 6 protein; Chronic lymphocytic leukemia deletion region gene 6 protein homolog; CLL deletion region gene 6 protein; CLL deletion region gene 6 protein homolog; Clld6; PUR1; RGD1306437; SPRY domain containing 7; SPRY domain-containing protein 7; SPRYD7
Gene Symbols Spryd7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CLLD6/C13orf1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57213
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.