Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLLD8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987250
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7250 Supplier Bioss Supplier No. BS6414R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Proteins that contain a SET domain, such as SETDB2, modulate gene expression epigenetically through histone H3 methylation. SETDB2 is likely a histone H3 methyltransferase, as it contains both the active site and flanking cysteine residues required for catalytic activity (Zhang et al., 2003 ).

Specifications

Antigen CLLD8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SETDB2
Gene Accession No. Q96T68
Gene Alias bifurcated 2; C13orf4; Chronic lymphocytic leukemia deletion region gene 8 protein; CLLD8; CLLL8; CLLL8 protein; DKFZp586I0123; DKFZp761J1217; Gm293; Histone-lysine N-methyltransferase SETDB2; KMT1F; Lysine N-methyltransferase 1F; SET domain; SET domain bifurcated 2; SET domain, bifurcated 2; SETDB2; Setdb2-Phf11-1-Phf11-2 fusion
Gene Symbols SETDB2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CLLD8/SETDB2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83852
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.