Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLM-4 (CD300c) Rat anti-Mouse, Unconjugated, Clone: TX52, MilliporeSigma™

Catalog No. MABF23282MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF23282MI 25 μg
MABF23281MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF23282MI Supplier MilliporeSigma Supplier No. MABF232825UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CLM-4 (CD300c) Monoclonal antibody specifically detects CLM-4 (CD300c) in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLM-4 (CD300c)
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TX52
Conjugate Unconjugated
Gene Accession No. NP_598919
Gene Alias CMRF-35-like molecule 4;CLM-4;CD300C molecule 2;Dendritic cell-derived Ig-like receptor 1;DIgR1;Immunoglobulin superfamily member 7;IgSF7;Leukocyte mono-Ig-like receptor 2;Myeloid-associated immunoglobulin-like receptor 2;MAIR-2;MAIR-II
Gene Symbols Cd300c2, Cd300c, Clm4, Igsf7, Lmir2
Host Species Rat
Immunogen Ba/F3 cells transfected with mouse CMRF-35-like molecule 4 (CD300c).
Molecular Weight of Antigen ∼25 and 33 kDa observed; 25.21 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone TX52 is a rat monoclonal antibody that specifically detects murine CMRF-35-like molecule 4 (CD300c).
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.