Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLM-9 (CD300c) Rat anti-Mouse, Unconjugated, Clone: ZAQ5, MilliporeSigma™

Catalog No. MABT15112MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT15112MI 25 μL
MABT15111MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT15112MI Supplier MilliporeSigma Supplier No. MABT151125UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CLM-9 (CD300c) Monoclonal antibody specifically detects CLM-9 (CD300c) in Mouse samples. It is validated for ELISA, Electron Microscopy, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLM-9 (CD300c)
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZAQ5
Conjugate Unconjugated
Gene Accession No. NP_082263.2
Gene Alias CMRF35-like molecule 9;CD300 antigen-like family member G;Nepmucin;CD300g
Gene Symbols Cd300lg, Clm9, D11Ertd736e
Host Species Rat
Immunogen Extracellular domain of mouse CMRF35-like molecule 9 (CLM-9; Nepmucin) protein fused with the Fc portion of human IgG
Molecular Weight of Antigen 36.76 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone ZAQ5 is a rat monoclonal antibody that detects murine CMRF35-like molecule 9 (CLM-9).
Target Species Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.