Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACD Monoclonal Antibody (1C11-1A7)
GREENER_CHOICE

Catalog No. PIMA524560
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524560 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524560 Supplier Invitrogen™ Supplier No. MA524560
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MPGRCQSDAA MRVNGPASRA PAGWTSGSLH TGPRAGRPRA QARGVRGRGL LLRPRPAKEL PLPRKGGAWA PAGNPGPLHP LGVAVGMAGS GRLVLRPWIR ELILGSETPS SPRAGQLLEV LQDAEAAVAG PSHAPDTSDV GATLLVSDGT HSVRCLVTRE ALDTSDWEEK EFGFRGTEGR LLLLQDCGVH VQVAEGGAPA EFYLQVDRFS LLPTEQPRLR VPGCNQDLDV QKKLYDCLEE HLSESTSSNA GLSLSQLLDE MREDQEHQGA LVCLAESCLT LEGPCTAPPV THWAASRCKA TGEAVYTVPS SMLCISENDQ LILSSLGPCQ RTQGPELPPP DPALQDLSLT LIASPPSSPS SSGTPALPGH MSSEESGTSI SLLPALSLAA PDPGQRSSSQ PSPAICSAPA TLTPRSPHAS RTPSSPLQSC TPSLSPRSHV PSPHQALVTR PQKPSLEFKE FVGLPCKNRP PFPRTGATRG AQEPCSVWEP PKRHRDGSAF QYEYEPPCTS LCARVQAARL PPQLMAWALH FLMDAQPGSE PTPM.

This gene encodes a uridine 5' -monophosphate synthase. The encoded protein is a bifunctional enzyme that catalyzes the final two steps of the de novo pyrimidine biosynthetic pathway. The first reaction is carried out by the N-terminal enzyme orotate phosphoribosyltransferase which converts orotic acid to orotidine-5' -monophosphate. The terminal reaction is carried out by the C-terminal enzyme OMP decarboxylase which converts orotidine-5' -monophosphate to uridine monophosphate. Defects in this gene are the cause of hereditary orotic aciduria. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACD
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1C11-1A7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ACD
Gene Accession No. Q96AP0
Gene Alias Acd; ACD shelterin complex subunit and telomerase recruitment factor; ACD, shelterin complex subunit and telomerase recruitment factor; adrenocortical dysplasia; adrenocortical dysplasia homolog; adrenocortical dysplasia homolog (mouse); adrenocortical dysplasia protein; Adrenocortical dysplasia protein homolog; PIP1; POT1 and TIN2-interacting protein; PTOP; TIN2 interacting protein 1; TINT1; TPP 1; TPP I; TPP1; TPPI
Gene Symbols ACD
Host Species Mouse
Immunogen ACD (AAH16904, 1 a.a. ~ 544 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65057
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.