Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596372 Supplier Invitrogen™ Supplier No. PA596372
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GANISTWVYS SPGRHEGQEP FLQWLMLLSN ESALPHVHTV SYGDDEDSLS SAYIQRVNTE LMKAAARGLT LLFASGDSGA GCWSVSGRHQ FRPTFPASSP YVTTVGGTSF QEPFLITNEI VDYISGGGFS NVFPRPSYQE EAVTKFLSSS PHLPPSSYFN ASGRAYPDVA ALSDGYWVVS NRVPIPWVSG TSASTPVFGG ILSLINEHRI LSGRPPLGFL NPRLYQQHGA GLFDVTRGCH ESCLDEEVEG QGFCSGPGWD PVTGWGTPNF PALLKTLLNP; Positive Samples: BT-474, SW480; Cellular Location: Lysosome, Melanosome.

This gene encodes a member of the sedolisin family of serine proteases. The protease functions in the lysosome to cleave N-terminal tripeptides from substrates, and has weaker endopeptidase activity. It is synthesized as a catalytically-inactive enzyme which is activated and auto-proteolyzed upon acidification. Mutations in this gene result in late-infantile neuronal ceroid lipofuscinosis, which is associated with the failure to degrade specific neuropeptides and a subunit of ATP synthase in the lysosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TPP1
Gene Accession No. O14773, O89023
Gene Alias cell growth-inhibiting gene 1 protein; ceroid-lipofuscinosis, neuronal 2; CLN 2; CLN2; GIG 1; GIG1; growth-inhibiting protein 1; LPIC; lysosomal pepstatin insensitive protease; lysosomal pepstatin-insensitive protease; MGC21297; SCAR7; TPP 1; TPP I; Tpp1; TPP-1; TPPI; TPP-I; Tripeptidyl aminopeptidase; tripeptidyl peptidase 1; tripeptidyl peptidase I; tripeptidyl peptidase-I; tripeptidyl-peptidase 1; Tripeptidyl-peptidase I; UNQ267/PRO304
Gene Symbols TPP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 284-563 of human TPP1 (NP_0003823).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1200, 12751
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.