Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLN2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547275
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the sedolisin family of serine proteases. The protease functions in the lysosome to cleave N-terminal tripeptides from substrates, and has weaker endopeptidase activity. It is synthesized as a catalytically-inactive enzyme which is activated and auto-proteolyzed upon acidification. Mutations in this gene result in late-infantile neuronal ceroid lipofuscinosis, which is associated with the failure to degrade specific neuropeptides and a subunit of ATP synthase in the lysosome.
Specifications
CLN2 | |
Polyclonal | |
Unconjugated | |
TPP1 | |
cell growth-inhibiting gene 1 protein; ceroid-lipofuscinosis, neuronal 2; CLN 2; CLN2; GIG 1; GIG1; growth-inhibiting protein 1; LPIC; lysosomal pepstatin insensitive protease; lysosomal pepstatin-insensitive protease; MGC21297; SCAR7; TPP 1; TPP I; Tpp1; TPP-1; TPPI; TPP-I; Tripeptidyl aminopeptidase; tripeptidyl peptidase 1; tripeptidyl peptidase I; tripeptidyl peptidase-I; tripeptidyl-peptidase 1; Tripeptidyl-peptidase I; UNQ267/PRO304 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
1200 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O14773 | |
TPP1 | |
Mouse myeloma cell line NS0-derived recombinant human Tripeptidyl-PeptidaseI/TPP1 Ser20-Pro563. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction