Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLN3 Polyclonal Antibody
CLN3 Polyclonal Antibody
Supplier: Thermo Scientific PA564103
Description
CLN3 Polyclonal Antibody for IHC

Specifications
CLN3 | |
Polyclonal | |
Unconjugated | |
CLN3 | |
AI323623; batten disease protein; Battenin; BTS; ceroid lipofuscinosis, neuronal 3, juvenile (Batten, Spielmeyer-Vogt disease); ceroid-lipofuscinosis, neuronal 3; CLN 3; Cln3; CLN3, battenin; Cln3p; JNCL; MGC102840; Protein CLN3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1201 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13286 | |
CLN3 | |
Recombinant protein corresponding to Human CLN3. Recombinant protein control fragment (Product #RP-104248). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction