Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109553
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109553 Supplier Invitrogen™ Supplier No. PA5109553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGGCAGSRRR FSDSEGEETV PEPRLPLLDH QGAHWKNAVG FWLLGLCNNF SYVVMLSAAH DILSHKRTSG NQSHVDPGPT PIPHNSSSRF DCNSVSTAAV LLADILPTLV IKLLAPLGLH LLPYSPRVLV SGICAAGSFV LVAFSHSVGT SLCGVVFASI SSGLGEVTFL SLTAFYPRAV ISWWSSGTGG AGLLGALSYL GLTQAGLSPQ QTLLSMLGIP ALLLASYFLL LTSPEAQDPG GEEEAESAAR QPLIRTEAPE SKPGSSSSLS LRERWTVFKG.

CLN3 is a highly glycosylated, hydrophobic, 438-amino acid protein with 6 transmembrane domains. The CLN3 protein localizes to the lysosomal membrane and plays a role in lysosomal function. It may act as a chaperone involved in the folding and unfolding of other proteins, namely subunit C of the ATP synthase complex. Mutations in the CLN3 gene cause Batten disease, a recessively inherited neurodegenerative disorder of childhood caused by lysosomal accumulation of hydrophobic material, mainly ATP synthase subunit C. Batten disease is the most common form of a group of disorders known as neuronal ceroid lipofuscinoses (NCLs). Symptoms of Batten disease include progressive loss of vision, seizures, and psychomotor disturbances.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLN3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLN3
Gene Accession No. Q13286, Q61124
Gene Alias AI323623; batten disease protein; Battenin; BTS; ceroid lipofuscinosis, neuronal 3, juvenile (Batten, Spielmeyer-Vogt disease); ceroid-lipofuscinosis, neuronal 3; CLN 3; Cln3; CLN3, battenin; Cln3p; JNCL; MGC102840; Protein CLN3
Gene Symbols CLN3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CLN3 (NP_0010358971).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1201, 12752, 293485
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.