Learn More
Invitrogen™ CLN5 Polyclonal Antibody

Description
Immunogen sequence: TLTGKNYTME WYELFQLGNC TFPHLRPEMD APFWCNQGAA CFFEGIDDVH WKENGTLVQV ATISGNMFNQ MAKWVKQDNE TGIYYETWNV KASPEK Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 85%.
Specifications
Specifications
| Antigen | CLN5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CLN5 |
| Gene Accession No. | O75503 |
| Gene Alias | A730075N08Rik; ceroid-lipofuscinosis; Ceroid-lipofuscinosis neuronal protein 5; ceroid-lipofuscinosis neuronal protein 5 homolog; Ceroid-lipofuscinosis neuronal protein 5 homolog, secreted form; Ceroid-lipofuscinosis neuronal protein 5, secreted form; ceroid-lipofuscinosis, neuronal 5; CLN5; NCL; Protein CLN5 |
| Gene Symbols | CLN5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.