Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588837 Supplier Invitrogen™ Supplier No. PA588837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPSRRHWPVP YKRFDFRPKP DPYCQAKYTF CPTGSPIPVM EGDDDIEVFR LQAPVWEFKY GDLLGHLKIM HDAIGFRSTL TGKNYTMEWY ELFQLGNCTF PHLRPEMDAP FWCNQGAACF FEGIDDVHWK ENGTLVQVAT ISGNMFNQMA KWVKQDNETG IYYETWNVKA SPEKGAETWF DSYDCSKFVL RTFNKLAEFG AEFKNIETNY TRIFLYSGEP TYLGNETSVF GPTGNKTLGL AIKRFYYPFK PHLPTKEFLL SLLQIFDAVI VHKQFYLFYN FEYWFLPMKF PFIKITYEEI PLPIRNKTLS GL; Positive Samples: Mouse heart, Rat heart; Cellular Location: Lysosome.

This gene is one of eight which have been associated with neuronal ceroid lipofuscinoses (NCL). Also referred to as Batten disease, NCL comprises a class of autosomal recessive, neurodegenerative disorders affecting children. The genes responsible likely encode proteins involved in the degradation of post-translationally modified proteins in lysosomes. The primary defect in NCL disorders is thought to be associated with lysosomal storage function.[provided by RefSeq, Oct 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLN5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CLN5
Gene Accession No. O75503, Q3UMW8
Gene Alias A730075N08Rik; ceroid-lipofuscinosis; Ceroid-lipofuscinosis neuronal protein 5; ceroid-lipofuscinosis neuronal protein 5 homolog; Ceroid-lipofuscinosis neuronal protein 5 homolog, secreted form; Ceroid-lipofuscinosis neuronal protein 5, secreted form; ceroid-lipofuscinosis, neuronal 5; CLN5; NCL; Protein CLN5
Gene Symbols CLN5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 47-358 of human CLN5 (NP_006484.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1203, 211286, 306128
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.