Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLN6 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305809AF647
Description
CLN6 Polyclonal specifically detects CLN6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CLN6 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ceroid-lipofuscinosis neuronal protein 6, ceroid-lipofuscinosis, neuronal 6, late infantile, variant, FLJ20561, HsT18960, nclf, Protein CLN6 | |
Produced in rabbits immunized with E. coli-derived Human CLN6 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
54982 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction