Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLN6, Mouse anti-Human, Clone: N400/24 , Millipore Sigma™

Catalog No. MABN254825 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN254825 25 μg
MABN2548100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN254825 Supplier MilliporeSigma Supplier No. MABN254825UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CLN6, clone N400/24, Cat. No. MABN2548, is a highly specific mouse monoclonal antibody that targets CLN6 and has been tested for use in Immunohistochemistry (Paraffin).

Ceroid-lipofuscinosis neuronal protein 6 (UniProt: Q9NWW5; also known as CLN6) is encoded by the CLN6 gene (Gene ID: 54982) in human. CLN6 is a multi-pass membrane protein that is localized to endoplasmic reticulum and contributes to lysosomal function and viability of neurons. It has no homology with known proteins or functional domains, however it is highly conserved across mammalian species. Mutations in CLN6 gene have been linked to ceroid lipofuscinosis, neuronal 6 and 4A that are characterized by intracellular accumulation of autofluorescent liposomal material, seizures, dementia, visual loss, and/or cerebral atrophy. Two isoforms of CLN6 have been described that are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLN6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone N400/24
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Ceroid-lipofuscinosis neuronal protein 6
Gene Symbols CLN6
Host Species Mouse
Immunogen Recombinant fragments corresponding to 54, 47, and 30 amino acids from non-helical regions from the N-terminal, internal, and C-terminal regions of human Ceroid-lipofuscinosis neuronal protein 6 (CLN6).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_060352.1
Test Specificity Clone N400/24 specifically detects human Ceroid-lipofuscinosis neuronal protein 6. It does not cross-react with other CLN proteins.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.