Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLN6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550006
Description
The antibody detects endogenous levels of total CLN6 protein.
CLN6, a 311-amino acid protein, has 7 predicted transmembrane domains and is conserved across vertebrates. The CLN6 protein localizes to the endoplasmic reticulum but contributes to lysosomal function. Mutations in the CLN6 gene cause variant late-onset infantile neuronal ceroid lipofuscinosis (vLINCL), a lysosomal storage disorder marked by progressive mental deterioration and blindness; part of a group of severe inherited neurodegenerative disorders affecting children wherein lysosomes accumulate storage material, causing the death of neurons. CLN6 is one of eight proteins, including CLN1-8, that are associated with NCL.
Specifications
CLN6 | |
Polyclonal | |
Unconjugated | |
CLN6 | |
1810065L06Rik; AW743417; ceroid-lipofuscinosis; ceroid-lipofuscinosis neuronal 6 late infantile; ceroid-lipofuscinosis neuronal protein 6; ceroid-lipofuscinosis, neuronal 6; ceroid-lipofuscinosis, neuronal 6, late infantile, variant; CLN4A; CLN6; D9Bwg1455e; HsT18960; nclf; neuronal ceroid lipofuscinosis; Protein CLN6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54982 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NWW5 | |
CLN6 | |
Synthesized peptide derived from internal of human CLN6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction