Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLN6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564260
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564260 Supplier Invitrogen™ Supplier No. PA564260
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WNDPVLRKKY PGVIYVPEPW AFYTLHVSSR Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 93%.

CLN6, a 311-amino acid protein, has 7 predicted transmembrane domains and is conserved across vertebrates. The CLN6 protein localizes to the endoplasmic reticulum but contributes to lysosomal function. Mutations in the CLN6 gene cause variant late-onset infantile neuronal ceroid lipofuscinosis (vLINCL), a lysosomal storage disorder marked by progressive mental deterioration and blindness; part of a group of severe inherited neurodegenerative disorders affecting children wherein lysosomes accumulate storage material, causing the death of neurons. CLN6 is one of eight proteins, including CLN1-8, that are associated with NCL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLN6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLN6
Gene Accession No. Q9NWW5
Gene Alias 1810065L06Rik; AW743417; ceroid-lipofuscinosis; ceroid-lipofuscinosis neuronal 6 late infantile; ceroid-lipofuscinosis neuronal protein 6; ceroid-lipofuscinosis, neuronal 6; ceroid-lipofuscinosis, neuronal 6, late infantile, variant; CLN4A; CLN6; D9Bwg1455e; HsT18960; nclf; neuronal ceroid lipofuscinosis; Protein CLN6
Gene Symbols CLN6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLN6. Recombinant protein control fragment (Product #RP-103621).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54982
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.