Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLN6 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8017R

Description
CLN6, a 311-amino acid protein, has 7 predicted transmembrane domains and is conserved across vertebrates. The CLN6 protein localizes to the endoplasmic reticulum but contributes to lysosomal function. Mutations in the CLN6 gene cause variant late-onset infantile neuronal ceroid lipofuscinosis (vLINCL), a lysosomal storage disorder marked by progressive mental deterioration and blindness; part of a group of severe inherited neurodegenerative disorders affecting children wherein lysosomes accumulate storage material, causing the death of neurons. CLN6 is one of eight proteins, including CLN1-8, that are associated with NCL.Specifications
CLN6 | |
Polyclonal | |
Unconjugated | |
CLN6 | |
1810065L06Rik; AW743417; ceroid-lipofuscinosis neuronal 6 late infantile; Ceroid-lipofuscinosis neuronal protein 6; ceroid-lipofuscinosis, neuronal 6; ceroid-lipofuscinosis, neuronal 6, late infantile, variant; CLN4A; CLN6; D9Bwg1455e; HsT18960; nclf; neuronal ceroid lipofuscinosis; Protein CLN6 | |
Rabbit | |
Protein A | |
RUO | |
54982 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9NWW5 | |
CLN6 | |
KLH conjugated synthetic peptide derived from human CLN6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction