Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLN8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107101 Supplier Invitrogen™ Supplier No. PA5107101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%;.

Could play a role in cell proliferation during neuronal differentiation and in protection against cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLN8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene CLN8
Gene Accession No. Q9UBY8
Gene Alias C8orf61; ceroid-lipofuscinosis, neuronal 8; ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation); Cln8; EPMR; mnd; motor neuron degeneration; protein CLN8
Gene Symbols CLN8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human CLN8 (aa 1-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2055
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.