Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLN8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549071
Description
CLN8 Polyclonal Antibody for Western Blot
Could play a role in cell proliferation during neuronal differentiation and in protection against cell death.
Specifications
CLN8 | |
Polyclonal | |
Unconjugated | |
CLN8 | |
C8orf61; ceroid-lipofuscinosis, neuronal 8; ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation); Cln8; EPMR; mnd; motor neuron degeneration; protein CLN8 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
2055 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9UBY8 | |
CLN8 | |
KLH conjugated synthetic peptide between 251-280 amino acids from the C-terminal region of human CLN8. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction