Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLN8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109554
Description
Immunogen sequence: MFHCRMVLTY HMWWVCFWHW DGLVSSLYLP HLTLFLVGLA LLTLIINPYW THKKTQQLLN PVDWNFAQPE AKSRPEGNGQ LLRKKRP.
Could play a role in cell proliferation during neuronal differentiation and in protection against cell death.
Specifications
| CLN8 | |
| Polyclonal | |
| Unconjugated | |
| CLN8 | |
| C8orf61; ceroid-lipofuscinosis, neuronal 8; ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation); Cln8; EPMR; mnd; motor neuron degeneration; protein CLN8 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2055 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.1 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9UBY8 | |
| CLN8 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction