Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLN8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113894 Supplier Invitrogen™ Supplier No. PA5113894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFGVQSTAAG LWALLGDPVL HADKARGQQN WCWFHITTAT GFFCFENVAV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 100%; Guinea Pig: 86%; Human: 100%; Mouse: 86%; Rabbit: 79%; Rat: 86%.

Could play a role in cell proliferation during neuronal differentiation and in protection against cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLN8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CLN8
Gene Accession No. Q9UBY8
Gene Alias C8orf61; ceroid-lipofuscinosis, neuronal 8; ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation); Cln8; EPMR; mnd; motor neuron degeneration; protein CLN8
Gene Symbols CLN8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human CLN8 (aa 72-121).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2055
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.