Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLN8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981608
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1608 Supplier Bioss Supplier No. BS11715R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Could play a role in cell proliferation during neuronal differentiation and in protection against cell death.

Specifications

Antigen CLN8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CLN8
Gene Accession No. Q9UBY8
Gene Alias C8orf61; ceroid-lipofuscinosis, neuronal 8; ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation); CLN8; EPMR; Protein CLN8
Gene Symbols CLN8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CLN8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2055
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.