Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLNK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557361
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557361 Supplier Invitrogen™ Supplier No. PA557361
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGNRKTTKEG SNDLKFQNFS LPKNRSWPRI NSATGQYQRM NKPLLDWERN FAAVLDGAKG HSDDDYDDPE LR Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 57%.

MIST is a member of the SLP76 family of adaptors. MIST plays a role in the regulation of immunoreceptor signaling, including PLC-gamma (PLCG1)-mediated B cell antigen receptor (BCR) signaling and FC-epsilon R1-mediated mast cell degranulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLNK
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLNK
Gene Accession No. Q7Z7G1
Gene Alias Clnk; cytokine dependent hematopoietic cell linker; cytokine-dependent hematopoietic cell linker; Mast cell immunoreceptor signal transducer; MIST; RGD1562417
Gene Symbols CLNK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLNK. Recombinant protein control fragment (Product #RP-96516).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116449
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.