Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLNS1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569809
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569809 Supplier Invitrogen™ Supplier No. PA569809
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 75%.

Putative Polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. PcG proteins are not required to initiate repression, but to maintain it during later stages of development. They probably act via methylation of histones, rendering chromatin heritably changed in its expressibility. Involved in transcriptional regulation mediated by ligand-bound nuclear hormone receptors, such as peroxisome proliferator-activated receptor gamma (PPARG). Acts as coactivator for PPARG and enhances its adipocyte differentiation-inducing activity; the function seems to involve differential recruitment of acetylated and methylated histone H3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLNS1A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CLNS1A
Gene Accession No. Q61189
Gene Alias 2610036D06Rik; 2610100O04Rik; ARPC1A; chloride channel; chloride channel current inducer; chloride channel regulator; chloride channel regulatory protein; chloride channel, nucleotide sensitive 1A; chloride channel, nucleotide-sensitive, 1A; chloride conductance regulatory protein ICln; Chloride ion current inducer protein; chloride nucleotide-sensitive channel 1A; ClCI; Clcni; Clns 1a; CLNS1A; CLNS1B; I(Cln); icln; icln protein; methylosome subunit pICln; pIcIn; pICln; Rcl1; Reticulocyte pICln; reticulocyte protein ICln; SOP2L; SOP2-like protein; swelling dependent chloride channel; swelling-induced chloride conductance regulatory protein; swelling-induced chloride conductance regulatory protein pIcIn
Gene Symbols CLNS1A
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse CLNS1A (aa 133-182).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12729
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.