Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Clobetasol Propionate Monoclonal Antibody (1B7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549727
Description
Antibody tested in indirect ELISA.
Clobetasol Propionate is a synthetic corticosteroid used to treat skin conditions such as eczema, psoriasis, and dermatitis. It works by reducing inflammation, itching, and redness in affected areas. It is available as a cream, ointment, or lotion and is applied topically to the affected area. Clobetasol Propionate should be used sparingly and only as directed by a healthcare professional, as prolonged use can lead to skin thinning and other adverse effects.
Specifications
Clobetasol Propionate | |
Monoclonal | |
1.75 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Conjugate of CBP hapten and KLH. | |
200 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
1B7 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction