Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLOCK Guinea Pig anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE259825UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2598-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2598-25UL Supplier MilliporeSigma Supplier No. ABE259825UL
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Anti-CLOCK is a highly specific guinea pig polyclonal antibody, that targets Circadian locomoter output cycles protein kaput and has been tested in Immunocytochemistry and ChIP-seq and Chromatin Immunoprecipitation (ChIP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLOCK
Applications ChIP Assay
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Clock
Host Species Guinea Pig
Immunogen KLH-conjugated linear peptide corresponding to 39 amino acids from the internal region of murine Circadian locomoter output cycles protein kaput (CLOCK).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This guinea pig polyclonal antibody detects Circadian locomoter output cycles protein kaput (CLOCK) in human and murine cells. It targets an epitope within 39 amino acids from the internal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.