Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLOCK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140985
This item is not returnable.
View return policy
Description
CLOCK Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.
Specifications
CLOCK | |
Polyclonal | |
Unconjugated | |
CLOCK | |
5330400M04Rik; bHLHe8; circadian locomoter output cycles kaput; circadian locomoter output cycles kaput protein; circadian locomoter output cycles protein kaput; circadian locomotor output cycles kaput; class E basic helix-loop-helix protein 8; CLOCK; clock circadian regulator; clock homolog; hCLOCK; I79_020252; KAT13D; KIAA0334; mCLOCK; rCLOCK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12753, 60447, 9575 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O08785, O15516, Q9WVS9 | |
CLOCK | |
Synthetic peptide within mouse CLOCK 829-855. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction