Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Clonidine hydrochloride
GREENER_CHOICESDP

Catalog No. 0690100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-901-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-901-00 Supplier Tocris Bioscience™ Supplier No. 0690/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

α2 agonist. Also I1 ligand

Prototypical I1 imidazoline receptor ligand. α2-adrenergic receptor agonist. Antihypertensive. Also thought to enhance autophagy; shown to increase LC3-II levels in PC12 cells.

Chemical Identifiers

CAS 4205-91-8
Molecular Formula C9H10Cl3N3

Specifications

Quantity 100 mg
Formula Weight 266.56
Purity >99%
Molecular Weight (g/mol) 266.55
Chemical Name or Material Clonidine hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.