Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (±)-Clopidogrel hydrochloride
GREENER_CHOICE

Catalog No. 249010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-901-0 10 mg
24-905-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-901-0 Supplier Tocris Bioscience™ Supplier No. 2490/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective P2Y12 antagonist

Selective, high affinity P2Y12 receptor antagonist (pKi = 6.9 for hP2Y12 receptor). Inhibits ADP-induced platelet aggregation; potent antithrombotic agent. Requires bioactivation, unlike PSB 0739 .

Active Enantiomer also available.

Chemical Identifiers

CAS 130209-90-4
Molecular Formula C16H17Cl2NO2S

Specifications

Quantity 10 mg
Formula Weight 358.28
Purity >99%
Molecular Weight (g/mol) 358.277
Chemical Name or Material (±)-Clopidogrel hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.