Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (+)-Clopidogrel hydrogen sulfate
GREENER_CHOICESDP

Catalog No. 182050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-205-0 50 mg
18-201-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-205-0 Supplier R&D Systems™ Supplier No. 1820/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective P2Y12 antagonist

Selective, high affinity P2Y12 receptor antagonist. Inhibits ADP-induced platelet aggregation and displays antithrombotic activity. Active enantiomer of (±)-clopidogrel hydrochloride (Cat. No. 2490).

Chemical Identifiers

CAS 120202-66-6
Molecular Formula C16H18ClNO6S2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 419.9
Purity >99%
Molecular Weight (g/mol) 419.89
Chemical Name or Material (+)-Clopidogrel hydrogen sulfate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.