Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clostridia Monoclonal Antibody (J44D)
GREENER_CHOICE

Catalog No. PIMA518193
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518193 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518193 Supplier Invitrogen™ Supplier No. MA518193
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specific for glutamate dehydrogenase of Clostridia species. It is reactive with recombinant GDH and native GDH from C. difficile cultures. Purity is >90%.

Clostridia are a highly polyphyletic class of Firmicutes, including Clostridium and other similar genera. Clostridia are distinguished from the Bacilli by lacking aerobic respiration. Clostridia are obligate anaerobes and oxygen is toxic to them. Diseases caused by Clostridia include botulism from C. botulinum as a result of exposure to botulinum toxin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridia
Applications ELISA
Classification Monoclonal
Clone J44D
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Clostridium
Host Species Mouse
Immunogen Clostridia recombinant protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.