Learn More
Invitrogen™ Clostridia Polyclonal Antibody, FITC

Description
The PA1-73169 antibody reacts with Clostridia species. PA1-73169 has been successfully used in Immunofluorescence applications. The PA1-73169 antibody was raised against an extract of Cl. perfingens, Cl. sordellii, Cl. novyi, Cl. septicum, Cl. Chauvoei.
Specifications
Specifications
| Antigen | Clostridia |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 |
| Gene Alias | Clostridium |
| Host Species | Rabbit |
| Immunogen | Extract of Cl. perfingens, Cl. sordellii, Cl. novyi, Cl. septicum, Cl. Chauvoei. |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.