Learn More
Clostridium botulinum D Toxoid Antibody - BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Clostridium botulinum D Toxoid |
| Applications | ELISA, Dot Blot |
| Classification | Polyclonal |
| Concentration | 28.9 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:100-1:2000, Dot Blot |
| Formulation | PBS with 0.09% Sodium Azide |
| Gene Alias | BoNT/D, Bontoxilysin D, BOTD, Botulinum neurotoxin type D |
| Host Species | Goat |
| Immunogen | Clostridium botulinum D toxoid, formalin fixed. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.