Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium botulinum D Toxoid Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017480 0.1 mL
NB10062469 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017480 Supplier Novus Biologicals Supplier No. NB100624690.1ML

Goat Polyclonal Antibody

Clostridium botulinum D Toxoid Polyclonal specifically detects Clostridium botulinum D Toxoid in Bacteria samples. It is validated for ELISA, Dot Blot.

Specifications

Antigen Clostridium botulinum D Toxoid
Applications Dot Blot, ELISA
Classification Polyclonal
Concentration 28.9 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Dot Blot
Formulation PBS with 0.09% Sodium Azide
Gene Alias BoNT/D, Bontoxilysin D, BOTD, Botulinum neurotoxin type D
Host Species Goat
Immunogen Clostridium botulinum D toxoid, formalin fixed.
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product recognizes C. botulinum toxoid type D, a secreted bacterial endopeptidase that inhibits neurotransmitter release.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.