Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clostridium botulinum HA-33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117583
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117583 Supplier Invitrogen™ Supplier No. PA5117583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Protects the structural integrity of the neurotoxin; may increase internalization of the neurotoxin into the bloodstream of the host. Involved in binding to the small intestine through interactions with glycolipids and glycoproteins containing sialic acid moieties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium botulinum HA-33
Applications Western Blot
Classification Polyclonal
Concentration 4.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HA-33
Gene Accession No. P46084
Gene Symbols HA-33
Host Species Rabbit
Immunogen Recombinant Clostridium botulinum Main hemagglutinin component protein (2-286aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3772939
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.